Learn More
Palintest™ Iron HR Test Kits
Product Code. 11923617
Click to view available options
Range:
0 to 10mg/L
Unit Size:
Pack of 50
Product Code. 11923617
Brand: Palintest™ PM156
Robust and reliable iron HR test kit for photometers.
  • Palintest Iron Test Reagent. 0 to 10 ppm, 50 Tests/Kit
  • The Palintest Iron HR test is based on a single tablet reagent containing an alkaline thioglycollate
  • The test is carried out simply by adding a tablet to a sample of the water under test
    • Iron (HR), Photometer Reagents, 0 - 10 mg/L Fe
Specifications
No. of Tests 50
Quantity 50 Pack
Range 0 to 10mg/L
Sufficient For 50 Tests
Type Iron HR Test Kit
compliance-icons compliance-icons
Product Identifier
  • Iron HR tablets
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H360FD-May damage fertility. May damage the unborn child.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P271-Use only outdoors or in a well-ventilated area.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P321-Specific treatment (see on this label).
  • P330-Rinse mouth.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : boric aicd, calciumthioglycolate