missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(+)-Irinotecan, TRC
Click to view available options
Quantity:
50 mg
250 mg
Unit Size:
250mg
50mg
Specifications
Specifications
| Chemical Name or Material | (+)-Irinotecan |
| CAS | 97682-44-5 |
| Molecular Formula | C33 H38 N4 O6 |
| Quantity | 250 mg |
| Synonym | [1,4'-Bipiperidine]-1'-carboxylic acid, (4S)-4,11-diethyl-3,4,12,14-tetrahydro-4-hydroxy-3,14-dioxo-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinolin-9-yl ester, [1,4'-Bipiperidine]-1'-carboxylic acid, 4,11-diethyl-3,4,12,14-tetrahydro-4-hydroxy-3,14-dioxo-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinolin-9-yl ester, (S)-, 1H-Pyrano[3',4':6,7]indolizino[1,2-b]quinoline, [1,4'-bipiperidine]-1'-carboxylic acid deriv., (+)-Irinotecan, Irinophore C, Irinotecan, Irinotecan lactone, Irinotecan mylan, Onivyde |
| InChI Formula | InChI=1S/C33H38N4O6/c1-3-22-23-16-21(43-32(40)36-14-10-20(11-15-36)35-12-6-5-7-13-35)8-9-27(23)34-29-24(22)18-37-28(29)17-26-25(30(37)38)19-42-31(39)33(26,41)4-2/h8-9,16-17,20,41H,3-7,10-15,18-19H2,1-2H3/t33-/m0/s1 |
| SMILES | CCc1c2CN3C(=O)C4=C(C=C3c2nc5ccc(OC(=O)N6CCC(CC6)N7CCCCC7)cc15)[C@@](O)(CC)C(=O)OC4 |
| Molecular Weight (g/mol) | 586.68 |
| Formula Weight | 586.2791 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction