missing translation for 'onlineSavingsMsg'
Learn More

Irinotecan hydrochloride trihydrate, MedChemExpress

Product Code. p-200100618
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16451226 200 mg 200mg
16427126 100 mg 100mg
16421816 500 mg 500mg
16413127 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16451226 Supplier Medchem Express Supplier No. HY16568

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Irinotecan hydrochloride trihydrate ((+)-Irinotecan hydrochloride trihydrate) is a topoisomerase I inhibitor with antitumor activity.

Irinotecan hydrochloride trihydrate ((+)-Irinotecan hydrochloride trihydrate) is a topoisomerase I inhibitor with antitumor activity.

Specifications

Chemical Name or Material Irinotecan hydrochloride trihydrate
CAS 136572-09-3
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C33H45ClN4O9
Quantity 200 mg
Synonym (+)-Irinotecan hydrochloride trihydrateCPT-11 hydrochloride trihydrate
Solubility Information DMSO : 50 mg/mL (73.84 mM; Need ultrasonic) ∣H2O : 1.52 mg/mL (2.24 mM; Need ultrasonic and warming)
SMILES [H]Cl.O=C(N1CCC(N2CCCCC2)CC1)OC3=CC=C4N=C5C(CN6C(C(COC([C@@]7(CC)O)=O)=C7C=C65)=O)=C(CC)C4=C3.O.O.O
Molecular Weight (g/mol) 677.18
Formula Weight 677.18
Percent Purity 98.44%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.