Learn More
Iridium Standard, ICP-MS Soectramass, 100 mg/L, Fisher Chemical™
BIOPHARMA
Product Code. 12977273 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Product Code. Quantity unitSize
12977273 100 mL 100mL
1 options
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 12977273
Brand: Fisher Chemical S 427 100
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Specifications
Concentration 100 mg/L
Solvent or Matrix Nitric Acid
Quantity 100 mL