missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ IREB2 (Human) Recombinant Protein
Human IREB2 full-length ORF ( AAH17880, 1 a.a. - 343 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00003658-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MDAPKAGYAFEYLIETLNDSSHKKFFDVSKLGTKYDVLPYSIRVLLEAAVRNCDGFLMKKEDVMNILDWKTKQSNVEVPFFPARVLLQDFTGIPAMVDFAAMREAVKTLGGDPEKVHPACPTDLTVDHSLQIDFSKCAIQNAPNPGGGDLQKAGKLSPLKVQPKKLPCRGQTTCRGSCDSGELGRNSGTFSSQIENTPILCPFHLQPVPEPETVLKNQEVEFGRNRERLQFFKWSSRVFKNVAVIPPGTGMAHQINLEYLSRVVFEEKDLLFPDSVVGTDSHITMVNGLGILGWGVGGIETEAVMLGLPVSLTLPEVVGCELTGSSNPFVTSIDVVLGITKVS
Specifications
AAH17880 | |
iron-responsive element binding protein 2 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
IREB2 | |
GST |
3658 | |
Wheat germ expression system | |
25 μg | |
ACO3, FLJ23381, IRP2, IRP2AD | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |