missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRAK-M Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA595154
This item is not returnable.
View return policy
Description
Synthetic peptide sequence: 349-364aa, KHLWYMPEEYIRQGRL. Reconstitution information: Add 0.2 mL of distilled water will yield a concentration of 500 μg/mL.
This gene encodes a member of the interleukin-1 receptor-associated kinase protein family. Members of this family are essential components of the Toll/IL-R immune signal transduction pathways. This protein is primarily expressed in monocytes and macrophages and functions as a negative regulator of Toll-like receptor signaling. Mutations in this gene are associated with a susceptibility to asthma. Alternate splicing results in multiple transcript variants.
Specifications
| IRAK-M | |
| Polyclonal | |
| 0.2mg sodium phosphate with 0.9MG NaCl, 0.9MG NaCl, 5MG BSA, 5MG BSA and 0.05MG sodium azide, 0.05MG sodium azide, 0.05MG thimerosal, 0.05MG thimerosal | |
| Q8K4B2, Q9Y616 | |
| Irak3 | |
| A synthetic peptide corresponding to a sequence at the C-terminus of mouse IRAKM, identical to the related rat sequence, and different from the related human sequence by one amino acid. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| IRAK3 | |
| Interleukin-1 receptor-associated kinase 3; IRAK-3; 2.7.11.1; IL-1 receptor-associated kinase M; IRAK-M; Irak3 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 11213, 314870, 73914 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction