missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IQCK Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25740-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene belongs to the IQ motif-containing family of proteins. The IQ motif serves as a binding site for different EF-hand proteins such as calmodulin. This gene was identified as a potential candidate gene for obsessive-compulsive disorder in a genome-wide association study. Alternative splicing of this gene results in multiple transcript variants.Specifications
| IQCK | |
| Polyclonal | |
| Unconjugated | |
| IQCK | |
| IQ domain containing protein K, IQ motif containing K, IQCK | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 124152, 434232 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8N0W5 | |
| Iqck | |
| IQCK Fusion Protein Ag22680 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction