Learn More
Thermo Scientific™ IPTG, dioxane-free
Product Code. p-4530700 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 g
5 g
5 x 5 g
Unit Size:
1g
25g
5g
Product Code. Quantity unitSize
10213090 1 g 1g
10725471 5 g 5g
10849040 5 x 5 g 25g
3 options
Product Code. 10213090
Brand: Thermo Scientific™ R0391
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Induce the expression of genes under the control of lac operon in cloning and protein expression procedures.
IPTG (isopropyl-beta-D-thiogalactopyranoside) is a highly stable synthetic analog of lactose. It inactivates the lac repressor and induces synthesis of beta-galactosidase, an enzyme that promotes lactose utilization. IPTG is used to induce expression of cloned genes under control of the lac operon . It is used in conjunction with X-Gal (#R0941) to determine the lac phenotype in blue/white colony screening.
It inactivates the lac repressor and induces synthesis of beta-galactosidase, an enzyme that promotes lactose utilization. IPTG is used to induce expression of cloned genes under control of the lac operon. It is used in conjunction with X-Gal to determine the lac phenotype in blue/white colony screening.
Ready-to-use 100mM IPTG aqueous solution is non-toxic, stable, 0.22μm membrane-filtered, formulated for direct use in blue/white colony screening.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) direct use in blue/white colony screening.
Product Type IPTG, dioxane-free
Quantity 1 g