Learn More
Invitrogen™ iPrep™ Cartridge Rack
Beskrivelse
- The iPrep™ Cartridge Rack is a stainless steel, alluminium alloy rack that holds the pre-filled carteridges during purification.
- The rack is designed to fit onto the plaform in only one orientation (indicated with an arrow).
- The racks dimensions are 10.8 inches (l) x 5.5 inches (w) x 2.3 inches (d).
- Each iPrep Instrument includes 1 iPRep Carteridge Rack. Additional racks can be ordered for faster instrument set up.
Tekniske data
Tekniske data
| Content And Storage | Store at room temperature. |
| For Use With (Equipment) | iPrep™ |
| Product Line | iPrep |
| Quantity | 1 rack (s) |
| Shipping Condition | Room Temperature |
Frequently Asked Questions (FAQs)
Do you still offer the iPrep Purification Instrument?
The iPrep Purification Instrument has been discontinued but we still offer iPrep purification kits and pre-programmed iPrep protocol cards to go along with the instrument.
For Research Use Only. Not for use in diagnostic procedures.
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?