Learn More
IPPK Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12603-1-AP-20UL
 View more versions of this product
Product Code. 16850404
  • 1710.33 NOK / 20µL
Description
The protein encoded by this gene is a kinase that phosphorylates position 2 of inositol-1, 3, 4, 5, 6-pentakisphosphate to form inositol-1, 2, 3, 4, 5, 6-hexakisphosphate (InsP6). InsP6 has a variety of functions, including stimulation of DNA repair, endocytosis, and mRNA export.
Specifications
Specifications
IPPK
Polyclonal
Unconjugated
IPPK
C9orf12, Ins(1,3,4,5,6)P5 2 kinase, InsP5 2 kinase, INSP5K2, IP5K, IPK1, IPK1 homolog, IPPK, KIAA0699
Rabbit
Antigen Affinity Chromatography
RUO
306808, 64768, 75678
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q5PXE9, Q6P1C1, Q9H8X2
IPPK
IPPK Fusion Protein Ag3278
20 μL
Primary
Mouse, Human, Rat