Learn More
IPO11 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Product Code. p-7205948 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16853024 20 μL 20µL
16843024 150 μL 150µL
2 options
Product Code. 16853024
Brand: Proteintech 144031AP20UL
Rabbit Polyclonal Antibody
This is a component of the ubiquinol-cytochrome c reductase complex (complex III or cytochrome b-c1 complex), which is part of the mitochondrial respiratory chain. The core protein 2 is required for the assembly of the complex.
Specifications
Antigen IPO11
Applications Immunoprecipitation, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Concentration 0.29 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene IPO11
Gene Accession No. Q8K2V6, Q9UI26
Gene Alias Imp11, importin 11, IPO11, Ran binding protein 11, RanBP11
Gene Symbols IPO11
Host Species Rabbit
Immunogen IPO11 Fusion Protein Ag5659
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 310056, 51194, 76582
Target Species Mouse, Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG