Learn More
Brady™ IP™ 6400 Series Thermal Transfer Printer Ribbon
Product Code. 10106074 Shop All Brady Products
Change view
Click to view available options
For Use With (Equipment):
BRADY IP Series, BradyPrinter i5100
Dimensions (L x W):
984 ft. x 1.5 in.
984 ft. x 2.36 in.
984 ft. x 3.27 in.
984 ft. x 4.3 in.
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Dimensions (L x W) Unit Size
10106074 BRADY IP Series, BradyPrinter i5100 984 ft. x 2.36 in. Each
11533854 BRADY IP Series, BradyPrinter i5100 984 ft. x 3.27 in. Each
12886816 BRADY IP Series, BradyPrinter i5100 984 ft. x 4.3 in. Each
12709336 BRADY IP Series, BradyPrinter i5100 984 ft. x 1.5 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10106074 Supplier Brady™ Supplier No. 066236
  • ONLINE EXCLUSIVE PRICE
    1980.00 NOK
    1908.80 NOK / Pack of 1
    Save 71.20 NOK
    3% Off
View all promotions
In Stock
High density ribbon with excellent solvent, high heat and super scratch resistance
  • Resin Formulation
  • Black color
Compatible with wide range of printers:
  • IP300, IP600 (F cat.11-878-501)
  • BBP72, BBP81, Brady 200MVP Plus, Brady 2461, Brady 300MVP Plus, Brady 300X-Plus II, Brady 3481, Brady 360X-Plus II, Brady 600X-Plus II, Brady 6441, Bradyprinter PR Plus, Bradyprinter PR Plus 600, PAM 3000, Tagus T200, Tagus T300, Thermal Transfer Printers (F cat. 22-500-855)
  • BBP11 (F cat.11-878-521)
Specifications
Certifications/Compliance RoHS 2002/95/EC
Description Black 6400 Series Thermal Transfer Printer Ribbon - Brady IP Printer Enabled
Adhesive Type Non-adhesive
Color Black
Finish Gloss
Format Label
Material Resin
Printable Sides Ink-side out
Print Technology None
Product Type Thermal Transfer Printer Label
Sterility Non-sterile
For Use With (Equipment) BRADY IP Series, BradyPrinter i5100
Quantity 1 Ea.
Size (English) 984 ft. x 2.36 in.
Thickness (English) 0.00018 in.
Thickness (Metric) 0.0045 mm
Dimensions (L x W) 984 ft. x 2.36 in.
Length (English) 984 ft.
Length (Metric) 300 m
Width (English) 2.36 in.
Width (Metric) 60 mm
Diameter (English) 1 in.
Diameter (Metric) 2.54 cm
Alternate Part Number 066236