Learn More
IOX2, MedChemExpress
Product Code. 16432647
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Product Code. 16432647
Brand: Medchem Express HY15468
MedChemExpress IOX2 is a specific prolyl hydroxylase-2 (PHD2) inhibitor with IC50 of 22 nM.
IOX2 is a specific prolyl hydroxylase-2 (PHD2) inhibitor with IC50 of 22 nM.
Specifications
Chemical Name or Material IOX2
Physical Form Solid
CAS 931398-72-0
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C19H16N2O5
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 25 mg/mL (70.95 mM; Need ultrasonic)
SMILES O=C(O)CNC(C1=C(O)C2=C(N(CC3=CC=CC=C3)C1=O)C=CC=C2)=O
Molecular Weight (g/mol) 352.34
Formula Weight 352.34
Percent Purity 98.33%
Grade Research
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.