Learn More
Thermo Scientific™ (IOPQ) Qualification and Performance Service
Product Code. 30049051 Shop All Thermo Scientific Products
Change view
Click to view available options
Product Type:
IQ/OQ/PQ Service
Unit Size:
1 unit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
30049051 IQ/OQ/PQ Service 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30049051 Supplier Thermo Scientific™ Supplier No. IOPQPCKE89003711
 Restricted Item
Estimated Shipment: 04-01-2027
Qualification service for New Heratherm Chamber 750 L (including documentation and 1 temp mapping).
  • IQ: Verifies that the installation site and equipment environment meet manufacturer-specified environmental requirements.
  • Verifies that the equipment, manuals, supplies and any other accessories arrived undamaged as specified in the sales order.
  • Verifies equipment and any other accessories are assembled and installed.
  • Configuration information for each system component gets recorded. Installation of Equipment is NOT Included.
  • OQ: Verifies important equipment functions and ensures that the equipment operates as expected by the manufacturer and in conformance to standards and requirements.
  • Key test: Door Ajar, Power Failure, Temperature Alerts PT: Temperature Mapping protocol performed at customer specified set point with provisions up to 15 measurement points, plus 1 ambient measurement point.
  • Temperature measurements will be performed in air or with simulated product loaded in chamber with a data collection interval of 1 minute for up to 24 hours, or configurable to customer needs.
  • Study requirements greater than 24 hours will be reviewed and quoted.
  • Temp Mapping service will include: (a) 1 open door temperature recovery test (b)1 power failure test (c) Min and max values along with averages (d) Temperature graph over a time period (e) Detailed reports will include raw probe data. All of this will be performed and delivered by a Thermo Fisher Qualified Engineer equipped with training certificates,temperature mapping data logger calibrated annually with metrology certificates.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Thermo Scientific New Heratherm Chamber 750 L
Product Type IQ/OQ/PQ Service