Learn More
Ion exchanger Dowex™ 50 WX-8, H+ form, EMD-Millipore
Specifications
Specifications
| CAS | 69011-20-7 |
| Molecular Formula | C28H30 |
| MDL Number | MFCD00132722 |
| Density | Density: 1.040g/cm3 (20°) |
| Synonym | porapak p, lewatit c267 ion exchange resin, ion exchange resin, benzene, diethenyl-, polymer with ethenylbenzene and ethenylethylbenzene, cation exchange resin, ethidium bromide adsorber, divinylbenzene, ethylvinylbenzene, vinylbenzene copolymer, ethenylbenzene, diethenylbenzene, ethylethenylbenzene polymer, ethylbenzene, ethylethenylbenzene, diethenylbenzene polymer, sulfonated, benzene, diethenyl-, polymer with ethenylbenzene and ethenylethylbenzene, sulfonated |
| Quantity | 250 g |
| InChI Key | NWUYHJFMYQTDRP-UHFFFAOYSA-N |
| SMILES | * |
| IUPAC Name | 1,2-diethenylbenzene; 1-ethenyl-2-ethylbenzene; ethenylbenzene |
| Molecular Weight (g/mol) | 0.00 |
| Show More |