Learn More
Ion Exchanger Amberlite™ IRA-402, Supelco™
Specifications
Specifications
| CAS | 52439-77-7 |
| MDL Number | MFCD00145564 |
| Density | 1.063 To 1.093 mg/L |
| Synonym | Ion exchanger Amberlite™ IRA-402, Ion exchanger Amberlite™ IRA-402 |
| Quantity | 500 mL |
| Chemical Name or Material | Ion Exchanger Amberlite™ IRA-402 |