missing translation for 'onlineSavingsMsg'
Learn More

Iodoethanol, TRC

Product Code. 30010650
Change view
Click to view available options
Quantity:
50 mg
250 mg
1 g
Unit Size:
1g
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010650 50 mg 50mg
30134282 250 mg 250mg
30137187 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010650 Supplier Toronto Research Chemicals Supplier No. TRCI70587550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 624-76-0
InChI Formula InChI=1S/C2H5IO/c3-1-2-4/h4H,1-2H2
IUPAC Name 2-iodoethanol
Molecular Formula C2 H5 I O
Molecular Weight (g/mol) 171.965
Chemical Name or Material Iodoethanol

Specifications

Quantity 50 mg
Synonym 2-Iodoethanol, 2-Hydroxy Iodide, 2-Iodoethan-1-ol, Ethylene Iodohydrin, NSC 85227
SMILES OCCI
Formula Weight 171.939
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.