Learn More
Iodoacetyl Chloride, TRC
Specifications
Specifications
| CAS | 38020-81-4 |
| Molecular Formula | C2H2ClIO |
| Quantity | 1 g |
| Synonym | 2-Iodoacetyl ChlorideI685625, 2-Iodoacetyl Chloride, (6CI,9CI)Iodoacetyl Chloride |
| InChI Formula | InChI=1S/C2H2ClIO/c3-2(5)1-4/h1H2 |
| SMILES | ClC(=O)CI |
| IUPAC Name | 2-iodoacetyl chloride |
| Molecular Weight (g/mol) | 204.39 |
| Formula Weight | 203.8839 |
| Chemical Name or Material | Iodoacetyl Chloride |