Learn More
Iodine Monobromide, Sigma-Aldrich™
Product Code. 15831151
Change view
Click to view available options
Quantity:
100 g
250 g
Unit Size:
100g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15831151 250 g 250g
15806660 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15831151 Supplier Sigma Aldrich Supplier No. 8207380250
 Restricted Item
Estimated Shipment: 18-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7789-33-5
Specifications
Boiling Point 116°C/1013 hPa (decomposition)
Melting Point 37°C to 40°C
Quantity 250 g
Density 4.41 g/cm3 at 20°C
MDL Number MFCD00011353
Synonym Iodine monobromide, Bromine iodide, Iodine monobromide
Molecular Weight (g/mol) 206.81
Grade Research
Chemical Name or Material Iodine Monobromide