missing translation for 'onlineSavingsMsg'
Learn More

Involucrin Antibody (SY5) - BSA Free, Novus Biologicals™

Product Code. p-200067828 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.10mg
Product Code. Quantity unitSize
18489258 0.1 mg 0.10mg
1 options
This item is not returnable. View return policy

Product Code. 18489258

Brand: Novus Biologicals NB1002727

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 1 publication

Involucrin Monoclonal specifically detects Involucrin in Human, Porcine, Canine, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, CyTOF-ready.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Involucrin
Applications Western Blot
Classification Monoclonal
Clone SY5
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:2000, Flow Cytometry 0.5 - 1.0 μg/106 cells, Immunohistochemistry 0.1-0.2 μg/mL, Immunocytochemistry/Immunofluorescence 1 - 2 μg/mL, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10-1:500, CyTOF-ready
Formulation PBS with 0.02% Sodium Azide
Gene Alias involucrin
Gene Symbols IVL
Host Species Mouse
Immunogen Purified involucrin from human keratinocytes.
Molecular Weight of Antigen 118 kDa
Purification Method Protein A purified
Quantity 0.1 mg
Research Discipline Extracellular Matrix
Primary or Secondary Primary
Gene ID (Entrez) 3713
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.