Learn More
Invitrogen™ X-Gal
Product Code. 11538746
Change view
Click to view available options
Quantity:
100 mg
1 g
Product Type:
X-Gal Substrate
Unit Size:
100mg
1g
Product Code. Quantity Product Type unitSize
11538746 100 mg X-Gal Substrate 100mg
10554973 1 g X-Gal Substrate 1g
2 options
Product Code. 11538746
Brand: Invitrogen™ 15520034
X-Gal (5-bromo-4-chloro-3-indolyl-β-D-galactoside) is a histochemical substrate for β-galactosidase. This substrate yields a blue precipitate upon hydrolysis, making it suitable for use in immunoblotting or immunocytochemical assays.
X-Gal (5-bromo-4-chloro-3-indolyl-β-D-galactoside) is a histochemical substrate for β-galactosidase. This substrate yields a blue precipitate upon hydrolysis, making it suitable for use in immunoblotting or immunocytochemical assays. Bacteria containing active β-galactosidase produce blue colonies when grown on plates containing X-Gal. X-Gal is supplied as white crystals and is soluble in dimethylformamide.
Primary applications
  • Blue-white screening—detect functional β-galactosidase (reporter gene expression)
  • Histochemical detection of β-galactosidase activity in fixed tissues
  • X-gal overlay assay
Find the media format that fits your needs
We offer many ready-to-use media formulations, such as LB Broth (10855001), Terrific Broth (A1374301) or S.O.C recovery medium (15544034).
We also offer IPTG (15529019) for blue-white screening.
Explore other media formats for bacterial growth.
Order Info
Approved for shipment at Room Temperature or on Wet Ice.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 5-bromo-4-chloro-3-indolyl-β-D-galactoside
Content And Storage Store in freezer (-5°C to -30°C).
Detection Method Colorimetric
Description X-Gal, Cell-Permeant
Format Bottle
For Use With (Application) Immunocytochemical Assays
Product Type X-Gal Substrate
Quantity 100 mg
Shipping Condition Approved for shipment at Room Temperature or on Wet Ice