Learn More
Invitrogen™ SYBR™ Green Nucleic Acid Gel Stain Starter Kit
Product Code. 10003232
Change view
Click to view available options
Quantity:
1 kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10003232 1 kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10003232 Supplier Invitrogen™ Supplier No. S7580
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Includes
Kit contains: 50μL of SYBR Green I nucleic acid gel stain (S-7567), 50μL of SYBR Green II RNA gel stain (S-7568) and a SYBR Green/Gold gel stain photographic filter (S-7569).
Everything needed for first time users to use SYBR gel stains
Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting
TRUSTED_SUSTAINABILITY
Specifications
Detection Location In-Gel Detection
Detection Method Fluorescence
Label or Dye SYBR Green II, SYBR Green I
Quantity 1 kit
Target Molecule RNA, DNA
Product Type Nucleic Acid Gel Stain Starter Kit