Learn More
Invitrogen™ Rhinohide™ Polyacrylamide Gel Strengthener Concentrate
Strengthens gels for easier handling and resistance to tearing
Brand: Invitrogen™ R33400
1013.38 NOK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Additional Details : Weight : 0.00525kg
Description
- Improves upon classic polyacrylamide gel technology, without adverse side effects
- Recommended for low-percentage gels, large-format gels and gels subject to multiple staining and handling steps
- Available in a kit (Cat. No. R-33410), which includes an acrylamide/bis-acrylamide mixture (37.5:1 ratio)
Gel Casting, Protein Gel Electrophoresis, Proteins, Expression, Isolation & Analysis
Order Info
Shipping Condition: Room Temperature
Specifications
Store at room temperature. | |
Rhinohide™ | |
Room Temperature |
Polyacrylamide Gel Strengthener Concentrate | |
200 mL |
For Research Use Only. Not for use in diagnostic procedures.