Learn More
Invitrogen™ pPICZα A, B, & C Pichia Vectors
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
Description
Includes
20μg each of lyophilized pPICZ A, B, and C is supplied
- Recombinant proteins are expressed as fusions to an N-terminal peptide encoding Saccharomyces cerevisiae '-factor secretion signal
- pPICα vectors contain AOX1 promoter
- All three reading frames (A, B, C versions) are provided to facilitate in-frame cloning with C-terminal peptide
- α-factor secretion signal for directing secreted expression of recombinant protein
- Zeocin resistance gene for selection in both E. coli and Pichia
- C-terminal peptide containing c-myc epitope and polyhistidine (6xHis) tag for detection and purification of recombinant fusion protein
Pichia pastoris Expression Systems, Protein Expression, Proteins, Expression, Isolation and Analysis, Yeast Expression
Specifications
Specifications
| Promoter | AOX1 |
| Product Type | Pichia Expression Vector |
| Content And Storage | 20 μg each of lyophilized pPICZ A, B, and C is supplied (Store at -20C) |
| Antibiotic Resistance Bacterial | Zeocin™ (ZeoR) |
| Shipping Condition | Room Temperature |
| Protein Tag | His Tag (6x), c-Myc Epitope Tag |
| Cloning Method | Restriction Enzyme/MCS |
| Quantity | 20 μg |
| Vector | pPIC |