Learn More
Invitrogen™ pcDNA™6/myc-His A, B, & C Mammalian Expression Vectors
Contain a strong promoter for high-level expression in mammalian cells
Brand: Invitrogen™ V22120
7063.20 NOK valid until 2024-06-28
Use promo code "21649" to get your promotional price.
Additional Details : Weight : 0.00525kg
Description
- Selection marker for generating stable cell lines
- Features epitope tag for easy detection with monoclonal antibody and rapid purification on nickel-chelating resin
- Three reading frames to simplify cloning in-frame with fusion tag
Constitutive Expression, Mammalian Expression, Protein Expression, Proteins, Expression, Isolation and Analysis
Specifications
Constitutive | |
Mammalian Expression Vector | |
Vectors (and appropriate expression control) are supplied supercoiled and lyophilized. Stored at -20°C. All vectors are guaranteed stable for 6 months when properly stored. | |
Restriction Enzyme/MCS | |
pcDNA™ |
CMV | |
Blasticidin | |
His Tag (6x), c-Myc Epitope Tag | |
pcDNA | |
Constitutive Expression |
For Research Use Only. Not for use in diagnostic procedures.