Learn More
Invitrogen™ pcDNA™3.1/His A, B, & C Mammalian Expression Vectors
Contain a strong promoter for high-level expression in mammalian cells
Brand: Invitrogen™ V38520
Additional Details : Weight : 0.02000kg
Includes: Vectors (and appropriate expression control) are supplied supercoiled and lyophilized.
Description
- Contains selection marker for generating stable cell lines and epitope tag for easy detection with monoclonal antibody and rapid purification on nickel-chelating resin
- Three reading frames to simplify cloning in-frame with fusion tag
Constitutive Expression, Mammalian Expression, Protein Expression, Proteins, Expression, Isolation and Analysis
Specifications
Constitutive | |
CMV | |
Geneticin™ (G-418) | |
EK (Enterokinase) Recognition Site | |
Restriction Enzyme/MCS | |
pcDNA™ |
Transfection | |
Mammalian Expression Vector | |
Vectors (and appropriate expression control) are supplied supercoiled and lyophilized. Stored at -20°C. All vectors are guaranteed stable for 6 months when properly stored. |
|
His Tag (6x), Xpress Epitope Tag | |
pcDNA | |
Constitutive Expression |
For Research Use Only. Not for use in diagnostic procedures.