Learn More
Invitrogen™ pAc5.1/V5-His A, B, & C Vectors
For high-level constitutive expression of recombinant proteins
Brand:  Invitrogen™ V411020
Additional Details : Weight : 0.02000kg
Product Code. 10717844
  • 8630.00 NOK / 1 set
Estimated Shipment: 19-06-2024
to see stock.
Includes: Contains 20μg each of a selection vector (pCoBlast or pCoHygro), pAc5.1/V5-His A, B, C, and pAc5.1/V5-His/lacZ.
Description
Description
  • The vector offers the strong, constitutive promoter from the Drosophila actin 5C gene
  • Small size (5.4 kb) to improve DNA yields and increase subcloning efficiency
  • C-terminal V5 epitope tag for rapid detection with Anti-V5 Antibody
  • C-terminal 6xHis tag for simple purification of recombinant fusion proteins using nickel-chelating resin
  • A set of three vectors-A, B, and C-is provided: Each vector has the multiple cloning site in a different reading frame relative to the coding sequence of the C-terminal tag
Constitutive Protein Expression in Insect Cells, High-level Protein Production in Insect Cells, Insect Expression, Protein Expression, Proteins, Expression, Isolation and Analysis
Specifications
Specifications
Lyophilized
Drosophila Expression Vector
His Tag (6x), V5 Epitope Tag
pAC
pAC5
Contains 20 µg each of lyophilized pAc5.1/V5-His A, B, C and pAc5.1/V5-His/LacZ.
Store at -20°C. All components are guaranteed stable for 6 months when properly stored.
Restriction Enzyme/MCS