Learn More
Invitrogen™ Oligo(dT)12-18 Primer
Product Code. 11513127
Change view
Click to view available options
Quantity:
50 μL
Unit Size:
25µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11513127 50 μL 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11513127 Supplier Invitrogen™ Supplier No. 18418012
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Oligo(dT)12-18 Primer is suitable for use in first-strand cDNA synthesis with reverse transcriptase.
Oligo(dT)12-18 Primer is suitable for use in first-strand cDNA synthesis with reverse transcriptase. The primer hybridizes to the poly(A) tail of mRNA. It is phosphorylated on the 5' end to facilitate cloning of cDNA.
Performance and quality testing: Performance is evaluated in a first-strand cDNA synthesis reaction
Order Info
Shipping Conditions: Approved for shipment on Wet or Dry Ice
TRUSTED_SUSTAINABILITY
Specifications
Product Type Primer
Content And Storage • Oligo(dT)12-18 Primer in DEPC-treated water (25 μg at 50 μg/μL)
Store at –20°C.
Guaranteed stable for 6 months when properly stored.
5'Primer Modification Phosphate
Primer Sequence 5'd PO4 [(T)12-18] 3'
Purification Method Gel-purified, Desalted
Shipping Condition Approved for shipment on Wet or Dry Ice
Concentration 50 μg/μL
Primer Oligo dT
Quantity 50 μL
For Use With (Application) First-strand cDNA Synthesis
Form Liquid