Learn More
Invitrogen™ NTP Mix (10 mM)
Product Code. 10505393
Change view
Click to view available options
Quantity:
20 μmol
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10505393 20 μmol Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27970” to get your promotional price
Product Code. 10505393 Supplier Invitrogen™ Supplier No. 18109017
Keep Your Science Moving with 3 for 2
Use promo code 27970
View all promotions
 Restricted Item
In Stock
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27970” to get your promotional price
ATP, CTP, GTP, UTP (nucleoside 5'-triphosphates), and 10 mM NTP Mix are suitable for use in in vitro transcription reactions with SP6, T3, and T7 RNA polymerases.
ATP, CTP, GTP, UTP (nucleoside 5´-triphosphates), and 10 mM NTP Mix are suitable for use in in vitro transcription reactions with SP6, T3, and T7 RNA polymerases for production of RNA probes, or RNA transcripts for translation in in vitro translation extracts. These nucleotides are supplied as convenient, ready-to-use solutions at a concentration of 10 mM in 1 mM Tris-HCl (pH 7.5).
Related products:
• 10 mM ATP (Cat. No. 18330019)
• 10 mM CTP (18331017)
• 10 mM GTP (18332015)
• 10 mM UTP (18333013)
Order Info
Approved for shipment on Wet or Dry Ice
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage NTP Mix, 10 mM (500 μL)
Store at –20°C.
Shipping Condition Approved for shipment on Wet or Dry Ice
Concentration 10 mM
Label or Dye Unlabeled
Quantity 20 μmol
For Use With (Application) in vitro Transcription
Form Liquid