Learn More
Invitrogen™ S.c. EasyComp™ Transformation Kit
Description
Includes
The S.c. EasyComp Transformation Kit provides enough reagents to prepare 6 batches of competent cells. Each batch is sufficient for 20 transformations.
- Cells can be made competent in less than 30 minutes
- Competent cell preparation and transformation solutions are optimized for use with S. Cerevisiae
- Provided ready-to-use and tested to ensure successful yeast transformation
- Competent cells can be frozen for future use
Cloning, Protein Expression, Proteins, Expression, Isolation and Analysis, S. cerevisiae Expression Systems, Transformation, Yeast Expression
Specifications
Specifications
| Product Type | Transformation Kit |
| Content And Storage | The S.c. EasyComp™ Transformation Kit provides enough reagents to prepare 6 batches of competent cells. Each batch is sufficient for 20 transformations. The kit should be stored at +4°C. All components are guaranteed stable for 6 months when properly stored. |
| Target Organism Class | S. cerevisiae |
| Quantity | 1 kit |
| Product Line | EasyComp |