Learn More
Invitrogen™ GTP
Description
ATP, CTP, GTP, UTP (nucleoside 5´-triphosphates), and 10 mM NTP Mix are suitable for use in in vitro transcription reactions with SP6, T3, and T7 RNA polymerases for production of RNA probes, or RNA transcripts for translation in in vitro translation extracts. These nucleotides are supplied as convenient, ready-to-use solutions at a concentration of 10 mM in 1 mM Tris-HCl (pH 7.5).
Related products
• 10 mM ATP (Cat. No. 18330019)
• 10 mM CTP (18331017)
• 10 mM UTP (18333013)
• 10 mM NTP Mix (18109017)
Order Info
Approved for shipment on Wet or Dry Ice
Specifications
Specifications
| Content And Storage | 10 mM GTP in Tris-HCl (250 μL) Store at –5 to –30°C. |
| Shipping Condition | Approved for shipment on Wet or Dry Ice |
| Concentration | 10 mM |
| Label or Dye | Unlabeled |
| Quantity | 500 μL |
| For Use With (Application) | in vitro Transcription |
| Form | Liquid |