Learn More
Invitrogen™ pLenti6/UbC/V5-DEST™ Gateway™ Vector
Product Code. 11510846
Change view
Click to view available options
Quantity:
6 μg
Unit Size:
6µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11510846 6 μg 6µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11510846 Supplier Invitrogen™ Supplier No. V49910
  • ONLINE EXCLUSIVE PRICE
    24430.00 NOK
    22475.60 NOK / 6µg
    Save 1954.40 NOK
    8% Off
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
Includes
pLenti6/UbC/V5-DEST 6μg, pLenti6/UbC/V5-GW/lacZ Control vector: 10μg; Stbl3 Chemically Competent E. coli(Box 2), Stbl3 Cells: 21 x 50μL, pUC19 control DNA (10pg/μL): 50μL, S.O.C. Medium: 6mL.
For lentiviral-based expression of a target gene in dividing and nondividing mammalian cells
  • The vector has the UbC promoter for driving constitutive but physiological levels of expression of the target gene and the blasticidin selection marker for stable selection
  • Suited to long-term experiments
  • Accurate titer of functional virus
  • V5 epitope tag at C terminus
Cloning, Constitutive Expression, Gateway Cloning, Mammalian Expression, Protein Expression, Proteins, Expression, Isolation and Analysis, Viral Gene Delivery
TRUSTED_SUSTAINABILITY
Specifications
Constitutive or Inducible System Constitutive
Delivery Type Lentiviral
Promoter UbC
Product Type Lentiviral Expression Vector
Selection Agent (Eukaryotic) Blasticidin
Content And Storage Vector Kit (Box 1)
pLenti6⁄UbC⁄V5-DEST™:6 μg, store at -20°C
pLenti6⁄UbC⁄V5-GW⁄lacZ Control vector: 10 ug, store at -20°C
One Shot™ Stbl3™ Chemically Competent E. coli(Box 2)
Stbl3™ Cells: 21 x 50 μL, store at -80°C
pUC19 control DNA (10 pg⁄μL): 50 μL, store at -80°C
S.O.C. Medium: 6 mL, store at -80°C
Protein Tag V5 Epitope Tag
Cloning Method Gateway
Quantity 6 μg
Vector pDEST, pLenti
Product Line Gateway, ViraPower
For Use With (Application) Viral Expression