Learn More
Invitrogen™ Gateway™ pDEST™17 Vector
Click to view available options
Quantity:
6 μg
Unit Size:
6µg
Description
Includes
pDEST 17 Vector is supplied in TE buffer (pH 8.0) at 150ng/μL.
Requires
Gateway entry clone, Gateway LR Clonase™ enzyme mix, and reaction buffer.
- Destination vectors have attR sites for recombination with any attL-flanked fragment, regardless of whether it is entry clone or UltimateRF Clone
Bacterial Expression,Cloning,Gateway Cloning,Protein Expression, Proteins, Expression, Isolation and Analysis
Specifications
Specifications
| Constitutive or Inducible System | Inducible |
| Promoter | T7 |
| Product Type | Expression Vector |
| Selection Agent (Eukaryotic) | None |
| Content And Storage | pDEST™ 17 Vector is supplied in TE buffer (pH 8.0) at 150ng/μl. Store at -20°C. Guaranteed for 6 months if stored properly. |
| Antibiotic Resistance Bacterial | Ampicillin (AmpR) |
| Protein Tag | His Tag (6x) |
| Cloning Method | Gateway |
| Quantity | 6 μg |
| Vector | pDEST, Gateway T7 Vectors |
| Show More |