Learn More
Invitrogen™ pBAD-DEST49 Gateway™ Destination Vector
Click to view available options
Quantity:
6 μg
Unit Size:
6µg
Description
Includes
The pBAD-DEST49 Gateway destination vector includes 6μg of vector.
- araBAD promoter for tightly regulated expression in E. coli
- HP-thioredoxin fusion for improved protein yield and solubility
- Enterokinase cleavage site for efficient cleavage of N-terminal fusion with EKMax™
- C-terminal V5 and 6xHis tags for efficient detection and purification of fusion proteins
- R sites for efficient recombination with any attL-flanked Gateway entry vector
Bacterial Expression/Cloning/Gateway Cloning/Protein Expression/Proteins, Expression, Isolation and Analysis
Specifications
Specifications
| Constitutive or Inducible System | Inducible |
| Inducing Agent | Arabinose |
| Promoter | araBAD |
| Product Type | Gateway System Destination Expression Vector |
| Selection Agent (Eukaryotic) | None |
| Content And Storage | The pBAD-DEST49 Gateway™ destination vector includes 6 μg of vector. Store at -20°C. The vector is guaranteed stable for 6 months when properly stored. |
| Antibiotic Resistance Bacterial | Ampicillin (AmpR) |
| Cleavage | EK (Enterokinase) Recognition Site |
| Protein Tag | His Tag (6x), V5 Epitope Tag, Thioredoxin |
| Cloning Method | Gateway |
| Show More |