missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dynabeads™ DETACHaBEAD™ Mouse CD4 Kit

Product Code. 10717513
Change view
Click to view available options
Quantity:
5 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10717513 5 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10717513 Supplier Invitrogen™ Supplier No. 12406D

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

DETACHaBEAD Mouse CD4, polyclonal Sheep anti-Mouse Fab to release mouse CD4+ T cells from Dynabeads Mouse CD4 (L3T4)

For detachment of viable mouse CD4+ T cells from Dynabeads; Mouse CD4 (L3T4)

  • Cell Type: T Cells (CD4
  • Isolation Method: Positive Isolation
  • Output Viability: >95%
  • Reactivity: Mouse
  • Sample Type: Blood, Lymph Node, Spleen
  • Starting Material (Cell No.): 1x107 PBMCs per isolation
  • Target Species: Mouse

Cell Analysis, Cell Isolation, Cell Isolation and Expansion, Mouse Cell Isolation

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in refrigerator (2–8°C).
For Use With (Application) Cell Analysis, Cell Isolation, Cell Expansion
No. of Cells 1 x 107 PBMCs per isolation
Purity or Quality Grade Research Grade
Quantity 5 mL
Sample Type Blood, Lymph Node, Spleen

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.