Learn More
Invitrogen™ Countess™ Power Cord with Plug Adapters
Click to view available options
:
Power Cord kit with Plug Adapters
Description
- Includes one power cord and four plug adapters that allows cord to be used in most countries
- Dimensions: 4 x 1m
Cell Counting, Viability and Cryopreservation
Specifications
Specifications
| Product Line | Countess |
| Content And Storage | Room temperature storage |
| Shipping Condition | Room Temperature |
| Dimensions | 4 x 1 meter |
| High-throughput Compatibility | Not High-throughput Compatible (Manual) |
| For Use With (Equipment) | Countess Automated Cell Counter |
| Product Type | Countess Power Cord with Plug Adapter |