Learn More
Invitrogen™ Countess™ Automated Cell Counter, with 101 Boxes of Slides
Product Code. 10185154
Click to view available options
Product Type:
Automated Cell Counter, 1 box of slides
Automated Cell Counter, 101 boxes of slides
Automated Cell Counter, 11 boxes of slides
Unit Size:
Each
Product Code. 10185154
Brand: Invitrogen™ C10310
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Includes
  • One Countess Automated Cell Counter, 101 boxes of 50 slides each (1050 total slides), 2 x 100mL bottles of Trypan blue stain, Power cord with 4 adaptors and Quick Reference Guide (C10311)
  • One Countess Automated Cell Counter, one box of 50 slides (100 total slides), 2 x 1mL vials of Trypan blue stain, power cord with 4 adaptors, and Quick Reference Guide (C10227)
  • One Countess Automated Cell Counter, 11 boxes of 50 slides each (550 total slides), 22 x 1mL vials of Trypan blue stain, Power cord with 4 adaptor and Quick Reference Guide (C10310)
Benchtop instrument for easy cell and viability counts in as little as 30 seconds
  • For verifying Number of Cells, Live Cells/mL, Dead Cells/mL, Cell Size
  • Compatible with Countess cell counting chamber slides
  • Dimensions: D x W x H: 20 x 27 x 19cm (7.8 x 10.6 x 7.4 in.)
Cell Counting, Viability and Cryopreservation
TRUSTED_SUSTAINABILITY
Specifications
Dimensions (D x W x H) 20 x 27 x 19cm
Voltage 100-240 VAC
Frequency 50/60 Hz
Product Type Automated Cell Counter, 11 boxes of slides
Includes One Countess Automated Cell Counter, 11 boxes of 50 slides each (550 total slides), 22 x 1mL vials of Trypan blue stain, Power cord with 4 adaptor and Quick Reference Guide