Learn More
Invitrogen™ ChargeSwitch™ gDNA Serum Kit, 0.2-1 mL
Kit includes necessary reagents
Brand:  Invitrogen™ CS11040
Additional Details : Weight : 0.20000kg
Product Code. 10707623
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Includes: ChargeSwitch Lysis Buffer (L19)<br />
ChargeSwitch Magnetic Beads<br />
Proteinase K (20 mg/mL in 50 mM Tris-HCl, pH 8.5, 5 mM CaCl2, 50% glycerol)<br />
ChargeSwitch Purification Buffer (N7)<br />
ChargeSwitch Wash Buffer (W12)<br />
ChargeSwitch Elution Buffer (E5; 10 mM Tris-HCl, pH 8.5)
Requires : Use of a magnetic accessory.
Description
Description
  • Genomic DNA for High-Performance PCR
  • Greater sensitivity and scalability than silica-based methods
  • Purifies DNA free of enzymatic inhibitors
DNA & RNA Purification & Analysis, DNA Extraction, Genomic DNA Purification, gDNA from Blood (Medium-Scale, 100 μl-1mL)
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
All components of the ChargeSwitch™ gDNA 1 ml Serum Kit are shipped at room temperature. Upon receipt, store the Proteinase K at 4“C. Store all other components at room temperature. All components are guaranteed stable for 6 months if stored properly.
Components:
ChargeSwitch™ Lysis Buffer (L19)
ChargeSwitch™ Magnetic Beads
Proteinase K (20 mg/ml in 50 mM Tris-HCl, pH 8.5, 5 mM CaCl2, 50% glycerol)
ChargeSwitch™ Purification Buffer (N7)
ChargeSwitch™ Wash Buffer (W12)
ChargeSwitch™ Elution Buffer (E5; 10 mM Tris-HCl, pH 8.5)
ChargeSwitch Chemistry
Real-Time Quantitative PCR (qPCR), PCR
High-throughput Compatible
100 μl - 1 ml Blood Samples ( Medium-Scale)
gDNA Serum Kit
Room Temperature
Kit
Blood
Fewer resources used and less waste, less hazardous
50 Preps
ChargeSwitch™
50 Preps
Genomic DNA