Learn More
Invitrogen™ BenchPro™ 4100 Western Card
Description
Includes
Box of 10 Cards and 10 membrane holders
Developed for immunodetection, the card holds the membrane to be processed in a Blot Holder. During operation, reagents are pumped from reagent bottles and vials located in the reagent tray, into the card and around the membrane. The order of reagents used and timing of protocol steps are based on the protocol selected.
Compatible for use with WP0001.
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Content And Storage | Box of 10 Cards and 10 membrane holders. Store at room temperature. |
| Capacity | 1 Membrane/Card |
| For Use With (Application) | Western Blot |
| For Use With (Equipment) | BenchPro™ 4100 Western Processing System, XCell SureLock™ Mini, iBlot™ |
| Product Line | BenchPro |
| Throughput | 1 Membrane/Card |
| Type | Card |
| For Use With | Compatible Compatible Products WP0001 |
| Quantity | 10 cards |
| Shipping Condition | Room Temperature |