Learn More
Gibco™ Bac-to-Bac™ HT Vector Kit
Product Code. 10022782
Change view
Click to view available options
Quantity:
1 kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10022782 1 kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10022782 Supplier Gibco™ Supplier No. 10584027
  • ONLINE EXCLUSIVE PRICE
    7100.00 NOK
    6532.00 NOK / 1 sets
    Save 568.00 NOK
    8% Off
View all promotions
 Restricted Item
Estimated Shipment: 11-12-2026
Includes
The Bac-to-Bac HT Vector Kit includes 10μg of pFastBac HT vector and pFastBac HT-CAT control vector.
Designed for use as part of Bac-to-Bac Baculovirus Expression System for expression and purification of histidine-tagged recombinant proteins in Sf9, Sf21, or High Five™ Cells following bacmid generation in E. coli
  • Strong polyhedrin promoter for protein expression
  • Three reading frames for simplified cloning
  • N-terminal 6 x His tag for simple purification of recombinant fusion proteins
  • TEV protease cleavage site for removal of histidine tag following protein purification
Baculovirus Production and Titering, Insect Expression, Protein Expression, Proteins, Expression, Isolation and Analysis
TRUSTED_SUSTAINABILITY
Specifications
Promoter Polyhedrin
Product Type Vector Kit
Content And Storage The Bac-to-Bac™ HT Vector Kit includes 10 μg of pFastBac™ HT vector and pFastBac HT-CAT control vector.
Store the vectors at -20°C. All components are guaranteed stable for 6 months when properly stored.
Cleavage TEV Protease Recognition Site
Protein Tag His Tag (6x)
Cloning Method Restriction Enzyme/MCS
Quantity 1 kit
Vector pFastBac
Product Line Bac-to-Bac