Learn More
Invitrogen™ Bac-N-Blue™ Transfection Kit
Product Code. 10646143
Change view
Click to view available options
Quantity:
5 transfections
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10646143 5 transfections Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10646143 Supplier Invitrogen™ Supplier No. K85501
  • ONLINE EXCLUSIVE PRICE
    10060.00 NOK
    9255.20 NOK / 5 reactions
    Save 804.80 NOK
    8% Off
 Restricted Item
Estimated Shipment: 11-12-2026
Includes
The Bac-N-Blue Transfection Kit contains 5 x 0.5μg vials of Bac-NBlue linear virus DNA and 125μL Cellfectin Reagent.
Specifically designed for recombination with pBlueBac vectors and pMelBac
  • Recombinant viruses have full-length, functional lacZ gene resulting in production of blue plaques for easy identification and purification
  • Bac-N-Blue DNA can be used with any polyhedrin promoter-based baculovirus transfer vector
  • Linearized at three sites, one of which is in gene that is essential for viral propagation: This leads to decrease in non-recombinant virus, making selection and purification of recombinant virus easy
  • In addition to viral DNA, Bac-N-BlueTransfection Kit contains Cellfectin Reagent for high transfection efficiency
Baculovirus Production and Titering, Insect Cell Transfection, Insect Expression, Protein Expression, Proteins, Expression, Isolation and Analysis
TRUSTED_SUSTAINABILITY
Specifications
Promoter Polyhedrin
Product Type Insect Cell Transfection Kit
Content And Storage The Bac-N-Blue™ Transfection Kit contains five 0.5 μg vials of Bac-NBlue™ linear virus DNA and 125 μl of Cellfectin™ Reagent.
Store at +4°C. All components are guaranteed stable for 6 months when properly stored.
Protein Tag Untagged
Cloning Method Restriction Enzyme/MCS
Quantity 5 transfections
Vector pBlueBac
Product Line Bac-N-Blue