Learn More
3M™ E-A-R™ E-A-Rsoft Yellow Earplugs
Product Code. 12364749 Shop All 3M Products
Change view
Click to view available options
Corded:
Corded
Uncorded
Unit Size:
1 pair
200 pairs
250 pairs
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Corded Unit Size
12364749 Uncorded 250 pairs
17237650 Uncorded 1 pair
12900365 Corded 200 pairs
11769305 Uncorded 250 pairs
11779305 Corded 200 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 12364749 Supplier 3M™ Supplier No. 7100111802
  • ONLINE EXCLUSIVE PRICE
    746.00 NOK
    433.15 NOK / 250 pairs
    Save 312.85 NOK
    41% Off
Estimated Shipment: 03-12-2026
Offers high attenuation and are made from a slow expanding, PU foam material. 3M E-A-R Polyurethane Earplug – 36dB.
  • Made from a slow expanding, PU foam material
  • Provides evenly distributed pressure, giving flexibility, and a good seal with optimum comfort.
  • Bright, neon color makes it appealing to wearers
TRUSTED_SUSTAINABILITY
Specifications
Color Yellow Neon
Certifications/Compliance CE Approved
For Use With (Application) For general industrial use
Quantity 250/Pk.
Noise Reduction Rating (NRR) 36 dB
Corded Uncorded
Product Type Ear Plugs
Material Polyurethane
Style Roll-down