Learn More
BINDER™ Interior Lighting
Product Code. 30238334 Shop All BINDER Products
Click to view available options
For Use With (Equipment):
KB 130, KB 260, KB 470
KB 1600
KB 65
KB 720, KB 1060
KB PRO 130/260/470
KB PRO 1600
KB PRO 260/470
KB PRO 720, KB PRO 1060
KBF 130, KBF 260, KBF 470
KBF 1600
KBF 720, KBF 1060
KBF PRO 1600
KBF PRO 720, KBF PRO 1060
Unit Size:
Each
Product Code. 30238334
Brand: BINDER™ 80122566
Only null left
Add to basket
Add to basket
Interior lighting with climate-resistant LEDs.
Interior lighting with climate-resistant LED lights, which turns on automatically when the unit's door is opened.
Specifications
For Use With (Equipment) KBF PRO 720, KBF PRO 1060
Material Glass
Type Lighting