Learn More
BINDER™ Interior Lighting
Product Code. 30238202 Shop All BINDER Products
Change view
Click to view available options
For Use With (Equipment):
KB 130, KB 260, KB 470
KB 1600
KB 65
KB 720, KB 1060
KB PRO 130/260/470
KB PRO 1600
KB PRO 260/470
KB PRO 720, KB PRO 1060
KBF 130, KBF 260, KBF 470
KBF 1600
KBF 720, KBF 1060
KBF PRO 1600
KBF PRO 720, KBF PRO 1060
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
30238202 KBF 1600 Each
30238222 KB 65 Each
30238266 KB 130, KB 260, KB 470 Each
30238246 KB PRO 130/260/470 Each
30238195 KBF 130, KBF 260, KBF 470 Each
30238198 KB PRO 260/470 Each
30238317 KB 720, KB 1060 Each
30238223 KB PRO 720, KB PRO 1060 Each
30238275 KBF 720, KBF 1060 Each
30238334 KBF PRO 720, KBF PRO 1060 Each
30238288 KB PRO 1600 Each
30238251 KBF PRO 1600 Each
30238305 KB 1600 Each
13 options
Product Code. 30238202
Brand: BINDER™ 80122569
Only null left
Add to basket
Add to basket
Interior lighting with climate-resistant LEDs.
Interior lighting with climate-resistant LED lights, which turns on automatically when the unit's door is opened.
Specifications
For Use With (Equipment) KBF 1600
Material Glass
Type Lighting