Learn More
Masterflex™ Interface cable RS-232 2.4m
Product Code. 12314558 Shop All Masterflex Products
Change view
Click to view available options
Length (Metric):
2.4 m
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
12314558 2.4 m Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12314558 Supplier Masterflex™ Supplier No. 2205054
  • ONLINE EXCLUSIVE PRICE
    366.00 NOK
    298.22 NOK / Pack of 1
    Save 67.78 NOK
    18% Off
Estimated Shipment: 22-01-2027
For software control to Masterflex™ pump
Specifications
For Use With (Equipment) Software control to pump
Length (Metric) 2.4 m
Includes RS232