missing translation for 'onlineSavingsMsg'
Learn More

Grant Instruments Interchangeable Head for Flat Universal Plate

Product Code. 17932701
Click to view available options
:
For Flat Universal Plate
This item is not returnable. View return policy

Product Code. 17932701

Brand: Grant Instruments H03

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Interchangeable Head for Controlled Rate Freeze Thaw (CRFT)

  • For use with Controlled Rate Freeze Thaw (CRFT)
  • Head for Flat Universal Plate
TRUSTED_SUSTAINABILITY

Specifications

Type Interchangeable Head
Exterior Finish Brushed Aluminum
Color Silver
For Use With (Application) Controlled Rate Freezing/Thawing
For Use With (Equipment) CRFT
Includes Packing Case
Defrost Manual, Software Controlled, or Automatic
Dimensions (L x W x H) Exterior 110 x 110 x 8.5 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.