Learn More
Grant Instruments™ Interchangeable Block for PCMT Thermoshaker
Product Code. 15159555
Change view
Click to view available options
No. of Places:
1
24
32
Holds:
0.2 mL/1.5 mL Microctubes
0.5/1.5 mL Microtubes
1.5 mL Microtubes
2.0 mL Microtubes
96-well Microplate (0.2 mL)
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Places Holds Unit Size
15159555 24 1.5 mL Microtubes Each
12337627 32 0.5/1.5 mL Microtubes Each
12347627 24 2.0 mL Microtubes Each
12909930 1 96-well Microplate (0.2 mL) Each
15137823 32 0.2 mL/1.5 mL Microctubes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 15159555 Supplier Grant Instruments™ Supplier No. HC24N
  • ONLINE EXCLUSIVE PRICE
    11385.00 NOK
    10521.47 NOK / Pack of 1
    Save 863.53 NOK
    7% Off
 Restricted Item
Estimated Shipment: 30-12-2026
Provides excellent uniformity and mixes evenly across the thermoshaker block. Grant Instruments™ Interchangeable Block for PCMT Thermoshaker is quick to reach specified mixing speed.
  • A block is required for use with PCMT Thermoshaker. Specify when ordering
  • Choice of 5 interchangeable blocks including a block suitable for microplates:
    • HC18 20 (20 x 0.5 ml and 12 x 1.5 ml microtubes)
    • HC32 (20 x 0.2 ml and 12 x 1.5 ml microtubes)
    • HC24N (24 x 1.5 ml microtubes)
    • HC24 (24 x 2 ml microtubes)
    • HC96 (96-well microplate for PCR, 0.2 ml) 
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) PCMT Thermoshaker
Holds 1.5 mL Microtubes
No. of Places 24
Temperature Range (Metric) 4°C to 100°C
Type Block
Temperature Uniformity ±0.1°C