Learn More
Grant Instruments Interchangeable Block for PCMT and PCMT PLUS Thermoshakers
Product Code. 30274325
Click to view available options
Holds:
20 x 10 mm Cuvette Tubes
20 x 12 mm Tubes
24 x 1.8 mL Cryovials
24 x 3.6 to 4.5 mL Cryovials
32 x 0.5 mL Microtubes
4 x 50 mL Tubes
8 x 15 mL Tubes
8 x 5 mL Tubes
Unit Size:
Each
Product Code. 30274325
Brand: Grant Instruments HC450
Only null left
Add to basket
Add to basket
Thermal blocks for use with PCMT and PCMT PLUS thermoshakers.
  • Block heating/cooling
  • Fits PCMT and PCMT PLUS
  • Max 1,400 rpm
Available to fit the following tubes:
  • 20 x 12 mm Tubes
  • 20 x 10 mm Cuvette Tubes
  • 24 x 3.6 to 4.5 mL Cryovials
  • 24 x 1.8 mL Cryovials
  • 32 x 0.5 mL Microtubes
  • 4 x 50 mL Tubes
  • 8 x 15 mL Tubes
  • 8 x 5 mL Tubes
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance CE
Color White, Blue
Autoclavable Not Autoclavable
Capacity (Metric) 50 to 500 mL
Chamber Material Aluminum
For Use With (Equipment) PCMT and PCMT PLUS
Holds 4 x 50 mL Tubes
Material Aluminum
No. of Places 4
Relative Humidity 80%
Temperature Range (Metric) 4°C to 80°C
Type Thermal Block
For Use With (Application) Thermal Mixing
No. of Blocks 1
Quantity 1 Ea.
Temperature Stability 0.1°C
Tube Size 50 mL
Warranty 2 Years
Dimensions (D x W x H) Interior 130 x 130 x 106 mm
Length (English) 5.12 in.
Length (Metric) 130 mm
Width (English) 5.12 in.
Width (Metric) 130 mm
Height (English) 4 in.
Height (Metric) 106 mm
Depth (English) 4 in.
Depth (English) Well 1.77 in.
Depth (Metric) 106 mm
Depth (Metric) Well 45 mm
Diameter (English) Base 5.12 in.
Diameter (English) Well 1.18 in.
Diameter (Metric) 130 mm
Diameter (Metric) Base 130 mm
Diameter (Metric) Well 30 mm
Shipping Weight (English) 2.2 lb.
Shipping Weight (Metric) 1 kg
Weight (English) 1.7 lb.
Weight (Metric) 0.8 kg