Learn More
Diversey™ IntelliCare Dispenser Manual
Product Code. p-7224790
Change view
Click to view available options
Color:
Black
White
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
17643105 Black Each
17653105 White Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17643105 Supplier Diversey™ Supplier No. D7524177
Only null left
Add to basket
Add to basket
The IntelliCare Manual Dispenser is a lock optional solution for hand care dispensing needs with a versatile, modern, and durable platform that is compatible with both liquid/gels and foam products.
  • State-of-the-art dispenser design with a large site window allows user content visibility without opening the dispenser
  • Ability to convert from keyed to keyless gives you the flexibility to choose your dispenser security levels
  • Ultimate versatility is achieved with one system that will dispense both liquid and foam products
  • Robust and reliable ABS-plastic, scratch and UV-resistant body with no dirt traps make for an easy maintenance and spotless appearance
  • Dose size adjustor allows you to vary the volume of soap dispensed based on end user needs
Specifications
Product Type Manual Dispenser
Color Black