missing translation for 'onlineSavingsMsg'
Learn More

Integrin alpha 4 beta 7 (LPAM-1) Monoclonal Antibody (DATK32 (DATK-32)), Functional Grade, eBioscience™, Invitrogen™

Product Code. p-7218523
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17101342 5 mg 5mg
17181332 25 mg 25mg
17191332 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17101342 Supplier Affymetrix eBioscience Supplier No. 16588738

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rat Monoclonal Antibody

The DATK32 MAb binds to a combinatorial epitope of mouse LPAM-1 (alpha4beta7), which is a heterodimer of the the 154 kDa alpha4 subunit and the 130 kDa beta7 subunit. In mouse, the beta7 subunit is found on the majority of mature lymphocytes and a small population of thymocytes and bone marrow cells. LPAM-1 is an integrin involved in the transendothelial migration of lymphocytes across high endothelial venules (HEV), and is know to bind several ligands including MAdCAM-1, VCAM-1 and fibronectin. Binding of the DATK32 MAb has been shown to induce aggregation of the CD8+ T cell lymphoma TK1, and block LPAM-1-mediated cell adhesion.

Integrin alpha 4 beta 7 (LPAM-1) Monoclonal antibody specifically detects Integrin alpha 4 beta 7 (LPAM-1) in Mouse samples. It is validated for Flow Cytometry, Functional Assay
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin alpha 4 beta 7 (LPAM-1)
Applications Flow Cytometry, Functional Assay
Classification Monoclonal
Clone DATK32 (DATK-32)
Concentration 1 mg/mL
Conjugate Functional Grade
Formulation PBS with no preservative; pH 7.2
Gene Itga4
Gene Accession No. P26011, Q00651
Gene Alias CD49 antigen-like family member D; CD49d; integrin alpha 4; integrin alpha-4; integrin alpha4B; Integrin alpha-IV; integrin beta 7; Integrin beta-7; Integrin beta-P; Itga4; Itga4B; Itgb7; LPAM subunit alpha; Ly69; Lymphocyte Peyer patch adhesion molecules subunit alpha; M290 IEL antigen; VLA; VLA 4; VLA-4 receptor, alpha 4 subunit; VLA4 subunit alpha; VLA-4 subunit alpha
Host Species Rat
Purification Method Affinity chromatography
Quantity 5 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16401, 16421
Target Species Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.